Gene Transcripts

Gene Symbol
Fermt2
Entrez Gene ID
218952
Gene Name
fermitin family homolog 2 (Drosophila)
Gene Aliases
AA960555, Kindlin-2, Mig2, Plekhc1
Chromosome Location
Chr. 14: 45458792 - 45530113 on Build GRCm38
UniGene ID
Mm.210018
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146054.2 NP_666166.2 12 1721
GenBank mRNA AK145030.1 - 12 1663
AK165608.1 - 12 1691
AK166552.1 - 12 1660
BC033436.1 - 12 1642 4950810
BC034168.1 - 3 375
CN716799.1 - 1 120

View Full Product Details