Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027905.2 | NP_082181.2 | 4, 5 | 1090 | |
GenBank mRNA | AK005048.1 | - | 4, 5 | 1090 | |
AK054432.1 | - | 4, 5 | 1052 | ||
AK088096.1 | - | 4, 5 | 1057 | ||
AK161059.1 | - | 4, 5 | 1041 | ||
BC048849.1 | - | 1, 2 | 113 | ||
BC117817.1 | - | 5, 6 | 605 | ||
BC117818.1 | - | 5, 6 | 638 | ||
BC141293.1 | - | 4, 5 | 1159 | 9055905 |