Gene Transcripts

Gene Symbol
Pi4ka
Entrez Gene ID
224020
Gene Name
phosphatidylinositol 4-kinase, catalytic, alpha polypeptide
Gene Aliases
Pik4ca
Chromosome Location
Chr. 16: 17280351 - 17406314 on Build GRCm38
UniGene ID
Mm.5718
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001983.2 NP_001001983.2 46 5257
GenBank mRNA AK136942.1 - 10 1044
BC022999.1 - 1 331
BC049252.1 - 6 595
BC075629.1 - 46 5257 6856662

View Full Product Details