Gene Transcripts

Gene Symbol
Fam13b
Entrez Gene ID
225358
Gene Name
family with sequence similarity 13, member B
Gene Aliases
2610024E20Rik, AW060714, AW546153
Chromosome Location
Chr. 18: 34440571 - 34523046 on Build GRCm38
UniGene ID
Mm.226899
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146084.1 NP_666196.1 14 2139
GenBank mRNA AK029766.1 - 15 2255
BC031465.1 - 14 2139 3981146

View Full Product Details