Gene Transcripts
- Gene Symbol
- Smc5
- Entrez Gene ID
- 226026
- Gene Name
- structural maintenance of chromosomes 5
- Gene Aliases
- MLZ-453, SMC-5, Smc5l1, mKIAA0594, mSMC5
- Chromosome Location
- Chr. 19: 23206441 - 23273908 on Build GRCm38
- UniGene ID
- Mm.23267
- Species
- Mus musculus
- Gene Symbol
- Naa15
- Entrez Gene ID
- 74838
- Gene Name
- N(alpha)-acetyltransferase 15, NatA auxiliary subunit
- Gene Aliases
- 5730450D16Rik, 6330400I15, ASTBDN, Narg1, Tbdn-1, mNAT1
- Chromosome Location
- Chr. 3: 51416016 - 51475985 on Build GRCm38
- UniGene ID
- Mm.275281
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF237622.1 |
- |
|
18 |
|
View Full Product Details