Gene Transcripts

Gene Symbol
PIK3CG
Entrez Gene ID
5294
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma
Gene Aliases
PI3CG, PI3K, PI3Kgamma, PIK3, p110gamma, p120-PI3K
Chromosome Location
Chr. 7: 106865278 - 106908978 on Build GRCh38
UniGene ID
Hs.32942
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282426.1 NP_001269355.1 4 2353
NM_001282427.1 NP_001269356.1 4 2277
NM_002649.3 NP_002640.2 4 2502
XM_005250443.3 XP_005250500.1 4 2370
XM_011516316.1 XP_011514618.1 4 2335
XM_011516317.1 XP_011514619.1 4 2335
XM_017012328.1 XP_016867817.1 4 2335
GenBank mRNA AF327656.1 - 3 2192
AK292684.1 - 4 2277
AK313753.1 - 4 2260
BC035683.1 - 4 2245 5749986
BX648341.1 - 4 2335
X83368.1 - 4 2512

View Full Product Details