Gene Transcripts

Gene Symbol
Ppapdc3
Entrez Gene ID
227721
Gene Name
phosphatidic acid phosphatase type 2 domain containing 3
Gene Aliases
D830019K17Rik, NET39
Chromosome Location
Chr. 2: 32095516 - 32110820 on Build GRCm38
UniGene ID
Mm.257236
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145521.3 NP_663496.1 2 1527
GenBank mRNA AK048016.1 - 3 1155
AK052883.1 - 2 1528
AK134638.1 - 2 1651
BC016136.1 - 2 1458 4910887

View Full Product Details