Gene Transcripts

Gene Symbol
Spg20
Entrez Gene ID
229285
Gene Name
spastic paraplegia 20, spartin (Troyer syndrome) homolog (human)
Gene Aliases
AI840044, C79168, SPARTIN, TAHCCP1, mKIAA0610
Chromosome Location
Chr. 3: 55112074 - 55137332 on Build GRCm38
UniGene ID
Mm.235523
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144987.1 NP_001138459.1 2 391
NM_001144988.1 NP_001138460.1 2 175
NM_144895.2 NP_659144.1 2 175
GenBank mRNA AK035081.1 - 1 34
AK087388.1 - 2 142
AK089946.1 - 2 175
AK129171.1 - 2 154
AK147760.1 - 2 392
AK159951.1 - 2 176
BC022921.1 - 2 143 4220506

View Full Product Details