Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145552.2 | NP_663527.2 | 7 | 840 | |
GenBank mRNA | AK037796.1 | - | 7 | 776 | |
AK077532.1 | - | 7 | 810 | ||
AK146404.1 | - | 7 | 837 | ||
BC003262.1 | - | 7 | 816 | 3501393 | |
U69600.1 | - | 7 | 735 |