Gene Transcripts

Gene Symbol
PLIN1
Entrez Gene ID
5346
Gene Name
perilipin 1
Gene Aliases
FPLD4, PERI, PLIN
Chromosome Location
Chr. 15: 89664365 - 89679417 on Build GRCh38
UniGene ID
Hs.103253
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145311.1 NP_001138783.1 8 1124
NM_002666.4 NP_002657.3 8 1195
XM_005254934.4 XP_005254991.1 8 1053
GenBank mRNA AB005293.1 - 8 1138
BC031084.1 - 8 1138 5284402
CB266260.1 - 1 12

View Full Product Details