Gene Transcripts

Gene Symbol
Wdtc1
Entrez Gene ID
230796
Gene Name
WD and tetratricopeptide repeats 1
Gene Aliases
Gm695, adipose, adp
Chromosome Location
Chr. 4: 133292466 - 133353476 on Build GRCm38
UniGene ID
Mm.226576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199306.4 NP_955010.1 10 1307
GenBank mRNA AK131700.1 - 12 1463
AK188298.1 - 3 161
AK213572.1 - 2 87
BC034833.1 - 3 230 5099225
BC048824.1 - 10 1101 4188979
BC057107.1 - 9 950 5697877

View Full Product Details