Gene Transcripts

Gene Symbol
Sel1l3
Entrez Gene ID
231238
Gene Name
sel-1 suppressor of lin-12-like 3 (C. elegans)
Gene Aliases
2310045A20Rik, AI429585, mKIAA0746
Chromosome Location
Chr. 5: 53107083 - 53213452 on Build GRCm38
UniGene ID
Mm.235020
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172710.3 NP_766298.2 15 2477
GenBank mRNA AK030487.1 - 15 2342
AK122362.1 - 15 2477
BC026655.1 - 3 200
BC094607.1 - 14 2022 6837048
BC117915.1 - 14 1978 40097358
BC117916.1 - 14 1980 40097359

View Full Product Details