Gene Transcripts

Gene Symbol
Samd4b
Entrez Gene ID
233033
Gene Name
sterile alpha motif domain containing 4B
Gene Aliases
BC042901
Chromosome Location
Chr. 7: 28399522 - 28436191 on Build GRCm38
UniGene ID
Mm.54120
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175021.3 NP_778186.2 3 907
GenBank mRNA AK185673.1 - 1 16
BC042901.1 - 3 960 5132736

View Full Product Details