Gene Transcripts

Gene Symbol
Mrgprb1
Entrez Gene ID
233231
Gene Name
MAS-related GPR, member B1
Gene Aliases
MrgB1
Chromosome Location
Chr. 7: 48444113 - 48456342 on Build GRCm38
UniGene ID
Mm.297430
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_205810.4 NP_991379.3 2 227
GenBank mRNA AK148044.1 - 2 223
AK148046.1 - 2 132
AK148052.1 - 2 131
AK148074.1 - 2 228
AK148108.1 - 2 132
AK148114.1 - 2 228
AK148166.1 - 2 132
AK148241.1 - 2 132
AK148253.1 - 2 223
AK148256.1 - 2 223
AK148273.1 - 2 200
AK148279.1 - 2 181
AK148322.1 - 2 165
AK148388.1 - 2 132
BC116395.1 - 1 53
BC116396.1 - 1 48

View Full Product Details