Gene Transcripts

Gene Symbol
Atmin
Entrez Gene ID
234776
Gene Name
ATM interactor
Gene Aliases
Asciz, gpg6, mKIAA0431
Chromosome Location
Chr. 8: 116943386 - 116960445 on Build GRCm38
UniGene ID
Mm.259736
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177700.4 NP_808368.3 2 383
GenBank mRNA AK122281.1 - 2 265
AX775769.1 - 2 383
BC027752.1 - 2 357
BC060631.1 - 2 368 6847850
CJ045227.1 - 2 386
CJ089804.1 - 3 305

View Full Product Details