Gene Transcripts

Gene Symbol
Hykk
Entrez Gene ID
235386
Gene Name
hydroxylysine kinase 1
Gene Aliases
Agphd1, C630028N24Rik
Chromosome Location
Chr. 9: 54917290 - 54949924 on Build GRCm38
UniGene ID
Mm.259334
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177351.4 NP_796325.2 4 1181
GenBank mRNA AK048771.1 - 3 1014
AK085527.1 - 4 1190
AK138851.1 - 4 1167
AK142509.1 - 3 3778
AK162425.1 - 4 1182
BC038618.1 - 4 1160 4221933

View Full Product Details