Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173413.3 | NP_775589.1 | 2 | 226 | |
GenBank mRNA | AB232601.1 | - | 2 | 154 | |
AK031251.1 | - | 2 | 173 | ||
AK051218.1 | - | 2 | 176 | ||
AK084650.1 | - | 2 | 176 | ||
AK155621.1 | - | 2 | 176 | ||
BC059208.1 | - | 2 | 208 | 6405462 | |
BC157974.1 | - | 2 | 174 | 9007296 | |
BY322741.1 | - | 2 | 227 |