Gene Transcripts

Gene Symbol
Mrgpra2b
Entrez Gene ID
235712
Gene Name
MAS-related GPR, member A2B
Gene Aliases
MrgA2, Mrgpra2, OTTMUSG00000028717
Chromosome Location
Chr. 7: 47463799 - 47489585 on Build GRCm38
UniGene ID
Mm.425459
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153101.3 NP_694741.2 2 217
GenBank mRNA AK156116.1 - 2 223
AY042192.1 - 191

Gene Symbol
Mrgpra2a
Entrez Gene ID
668727
Gene Name
MAS-related GPR, member A2A
Gene Aliases
Chromosome Location
Chr. 7: 47426325 - 47452167 on Build GRCm38
UniGene ID
Mm.431311
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172588.1 NP_001166059.1 192
GenBank mRNA BC064040.1 - 192

View Full Product Details