Gene Transcripts

Gene Symbol
Fam83a
Entrez Gene ID
239463
Gene Name
family with sequence similarity 83, member A
Gene Aliases
Chromosome Location
Chr. 15: 57985903 - 58010706 on Build GRCm38
UniGene ID
Mm.221241
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173862.2 NP_776287.2 2 791
GenBank mRNA BC030396.1 - 2 794

View Full Product Details