Gene Transcripts

Gene Symbol
Pdzrn4
Entrez Gene ID
239618
Gene Name
PDZ domain containing RING finger 4
Gene Aliases
1110017D07Rik, LNX4, SAMCAP3L, mKIAA3027
Chromosome Location
Chr. 15: 92396810 - 92771827 on Build GRCm38
UniGene ID
Mm.309826
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164593.1 NP_001158065.1 9 1552
NM_001164594.1 NP_001158066.1 7 989
GenBank mRNA AK135524.1 - 7 972
BC151034.1 - 9 1481 9087947

View Full Product Details