Gene Transcripts

Gene Symbol
Dusp27
Entrez Gene ID
240892
Gene Name
dual specificity phosphatase 27 (putative)
Gene Aliases
C130085G02Rik, Gm209
Chromosome Location
Chr. 1: 166098148 - 166127898 on Build GRCm38
UniGene ID
Mm.35315
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033344.3 NP_001028516.2 6 2869
NM_001160049.1 NP_001153521.1 6 2865
GenBank mRNA AK048603.1 - 6 2892
AK142263.1 - 6 2870
BC059034.1 - 2 2218
BC117936.1 - 6 2846 40097789

View Full Product Details