Gene Transcripts

Gene Symbol
Lrrc8a
Entrez Gene ID
241296
Gene Name
leucine rich repeat containing 8A
Gene Aliases
Lrrc8, mKIAA1437
Chromosome Location
Chr. 2: 30237769 - 30263790 on Build GRCm38
UniGene ID
Mm.389232
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177725.4 NP_808393.1 3 466
GenBank mRNA BQ444045.1 - 3 514
BQ745966.1 - 3 467

View Full Product Details