Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001024618.2 | NP_001019789.1 | 7 | 1191 | |
NM_001083919.1 | NP_001077388.1 | 7 | 1191 | ||
GenBank mRNA | AK040786.1 | - | 7 | 1192 | |
AY775570.1 | - | 6 | 1045 | ||
DQ011666.1 | - | 7 | 1192 | ||
EF119716.1 | - | 7 | 1191 | ||
EF122140.1 | - | 6 | 946 |