Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172856.3 | NP_766444.1 | 3 | 411 | |
GenBank mRNA | AK028849.1 | - | 3 | 412 | |
AK135798.1 | - | 3 | 513 | ||
AK151848.1 | - | 3 | 500 | ||
AK152417.1 | - | 3 | 481 | ||
BC057629.1 | - | 3 | 454 | 4224547 |