Gene Transcripts

Gene Symbol
Tspan18
Entrez Gene ID
241556
Gene Name
tetraspanin 18
Gene Aliases
2610042G18Rik, 6720430O15, BB226562
Chromosome Location
Chr. 2: 93201760 - 93334555 on Build GRCm38
UniGene ID
Mm.274159
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183180.2 NP_899003.1 7 750
GenBank mRNA AK032765.1 - 6 661
BC052169.1 - 7 713 6335221

View Full Product Details