Gene Transcripts

Gene Symbol
Lingo2
Entrez Gene ID
242384
Gene Name
leucine rich repeat and Ig domain containing 2
Gene Aliases
B230217C06Rik, Lern3, Lrrn6c
Chromosome Location
Chr. 4: 35706647 - 36952254 on Build GRCm38
UniGene ID
Mm.132507
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165999.1 NP_001159471.1 8 1571
NM_001166000.1 NP_001159472.1 9 1030
NM_001166001.1 NP_001159473.1 2 491
NM_175516.4 NP_780725.2 3 598
GenBank mRNA AK035778.1 - 3 540
AK045639.1 - 3 599
AK141568.1 - 8 1054
BC090619.1 - 9 958 6734654
BC137866.1 - 2 432 8860885
BC144985.1 - 2 432 9053533
BC145692.1 - 2 432 40130995

View Full Product Details