Gene Transcripts

Gene Symbol
Rab42
Entrez Gene ID
242681
Gene Name
RAB42, member RAS oncogene family
Gene Aliases
9530096D07Rik, Rab42-ps
Chromosome Location
Chr. 4: 132300438 - 132303403 on Build GRCm38
UniGene ID
Mm.331927
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081651.1 NP_001075120.1 2 353
GenBank mRNA AB232642.1 - 2 334
AK035722.1 - 1 698
BC132060.1 - 2 382 40130337
BC132062.1 - 2 382 40130339

View Full Product Details