Gene Transcripts
- Gene Symbol
- Sspo
- Entrez Gene ID
- 243369
- Gene Name
- SCO-spondin
- Gene Aliases
- C79529, Scospondin
- Chromosome Location
- Chr. 6: 48446243 - 48501262 on Build GRCm38
- UniGene ID
- Mm.25039
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173428.3 |
NP_775604.3
|
29 |
4534 |
|
GenBank mRNA |
AJ491857.1 |
- |
27 |
4159 |
|
View Full Product Details