Gene Transcripts

Gene Symbol
Chrm2
Entrez Gene ID
243764
Gene Name
cholinergic receptor, muscarinic 2, cardiac
Gene Aliases
AChR-M2, Chrm-2, M2
Chromosome Location
Chr. 6: 36387950 - 36528638 on Build GRCm38
UniGene ID
Mm.448632
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203491.3 NP_987076.2 3 873
GenBank mRNA BC019470.1 - 3 873

View Full Product Details