Gene Transcripts

Gene Symbol
Cnksr2
Entrez Gene ID
245684
Gene Name
connector enhancer of kinase suppressor of Ras 2
Gene Aliases
C030004A01, Cnk2, Ksr2, mKIAA0902
Chromosome Location
Chr. X: 157821573 - 158043313 on Build GRCm38
UniGene ID
Mm.197074
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177751.2 NP_808419.1 19 2355
GenBank mRNA AK122399.1 - 18 2279
BC043093.1 - 19 2355 6405389
BC060716.1 - 19 2355 6405389

View Full Product Details