Gene Transcripts

Gene Symbol
Prrt1
Entrez Gene ID
260297
Gene Name
proline-rich transmembrane protein 1
Gene Aliases
Bat-4, Bat4, D17H6S54E, DSPD1, G5b, NG5, ORF31
Chromosome Location
Chr. 17: 34629569 - 34633134 on Build GRCm38
UniGene ID
Mm.390965
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030890.1 NP_112152.1 4 868
GenBank mRNA BC115512.2 - 4 671
BC115513.2 - 4 671
BC128489.1 - 5 537

View Full Product Details