Gene Transcripts

Gene Symbol
Otos
Entrez Gene ID
260301
Gene Name
otospiralin
Gene Aliases
OTOSP
Chromosome Location
Chr. 1: 92644218 - 92648841 on Build GRCm38
UniGene ID
Mm.219545
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153114.2 NP_694754.1 5 470
GenBank mRNA AK083090.1 - 4 2758
AY078071.1 - 4 218
BC100323.1 - 5 470 30788363
BC132494.1 - 4 389 40130771
BC132498.1 - 4 389 40130775

View Full Product Details