Gene Transcripts

Gene Symbol
Creg2
Entrez Gene ID
263764
Gene Name
cellular repressor of E1A-stimulated genes 2
Gene Aliases
A830098L22Rik
Chromosome Location
Chr. 1: 39618406 - 39651510 on Build GRCm38
UniGene ID
Mm.122091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170597.4 NP_733485.1 4 998
GenBank mRNA AB046110.1 - 4 1316
AK034501.1 - 4 761
AK043871.1 - 4 763
AK044192.1 - 4 998
AK046245.1 - 4 785
AK048589.1 - 4 792
AK134713.1 - 4 769
AK134861.1 - 4 1059
BC034115.1 - 4 686
BC116790.1 - 4 891 40126109
BC116792.1 - 4 891 40126111

View Full Product Details