Gene Transcripts

Gene Symbol
Lrrc16b
Entrez Gene ID
268747
Gene Name
leucine rich repeat containing 16B
Gene Aliases
mFLJ00240
Chromosome Location
Chr. 14: 55490656 - 55508272 on Build GRCm38
UniGene ID
Mm.151577
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024645.1 NP_001019816.1 8 708
GenBank mRNA AK148355.1 - 8 731
BC082805.1 - 8 628
BC092223.1 - 8 626

View Full Product Details