Gene Transcripts

Gene Symbol
Clpx
Entrez Gene ID
270166
Gene Name
caseinolytic mitochondrial matrix peptidase chaperone subunit
Gene Aliases
AU014732, E330029I21
Chromosome Location
Chr. 9: 65294250 - 65330658 on Build GRCm38
UniGene ID
Mm.30088
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044389.2 NP_001037854.1 7 1168
NM_011802.3 NP_035932.2 8 1210
GenBank mRNA AF134983.1 - 8 1161
AJ276991.1 - 8 1092
AK087848.1 - 8 1164
BC061153.1 - 7 1126 6593118

View Full Product Details