Gene Transcripts

Gene Symbol
Emilin3
Entrez Gene ID
280635
Gene Name
elastin microfibril interfacer 3
Gene Aliases
1110013O17Rik, EMILIN-3, EMILIN-5, EMILIN-T, Emilin5
Chromosome Location
Chr. 2: 160906438 - 160912339 on Build GRCm38
UniGene ID
Mm.330747
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291145.1 NP_001278074.1 4 1529
NM_182840.2 NP_878260.1 4 1670
GenBank mRNA BC054738.1 - 4 1659
BC057619.1 - 4 1659 6402350

View Full Product Details