Gene Transcripts

Gene Symbol
Adgrl3
Entrez Gene ID
319387
Gene Name
adhesion G protein-coupled receptor L3
Gene Aliases
5430402I23Rik, CIRL-3, D130075K09Rik, Gm1379, LEC3, Lphn3, mKIAA0768
Chromosome Location
Chr. 5: 81021199 - 81827825 on Build GRCm38
UniGene ID
Mm.273631
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198702.2 NP_941991.1 9 1777
GenBank mRNA AK051766.1 - 8 1455
AK140842.1 - 7 1616
AK147374.1 - 1872
AK155931.1 - 8 2097
AK157740.1 - 8 1356
BC058992.1 - 1 222
BC088989.1 - 9 1777 5705210
BC094668.1 - 7 1586

View Full Product Details