Gene Transcripts

Gene Symbol
Agmo
Entrez Gene ID
319660
Gene Name
alkylglycerol monooxygenase
Gene Aliases
A530016O06Rik, AI790538, Tmem195
Chromosome Location
Chr. 12: 37241629 - 37581932 on Build GRCm38
UniGene ID
Mm.485540
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178767.5 NP_848882.2 1 460
GenBank mRNA AK039191.1 - 1 31
AK040702.1 - 1 461
AK050246.1 - 1 321
AK163155.1 - 1 241
BC038044.1 - 1 360

View Full Product Details