Gene Transcripts

Gene Symbol
Arhgef37
Entrez Gene ID
328967
Gene Name
Rho guanine nucleotide exchange factor (GEF) 37
Gene Aliases
4933429F08Rik
Chromosome Location
Chr. 18: 61493625 - 61537044 on Build GRCm38
UniGene ID
Mm.488098
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177828.4 NP_808496.2 5 783
GenBank mRNA AK030242.1 - 1 610

View Full Product Details