Gene Transcripts

Gene Symbol
Snapc5
Entrez Gene ID
330959
Gene Name
small nuclear RNA activating complex, polypeptide 5
Gene Aliases
2010103A03Rik
Chromosome Location
Chr. 9: 64179297 - 64182688 on Build GRCm38
UniGene ID
Mm.24082
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183316.2 NP_899139.2 3 239
GenBank mRNA AK007634.1 - 2 75
AK010023.1 - 2 75
BC028529.1 - 3 228 1381169
CJ165571.1 - 3 240

View Full Product Details