Gene Transcripts
- Gene Symbol
- RAB5B
- Entrez Gene ID
- 5869
- Gene Name
- RAB5B, member RAS oncogene family
- Gene Aliases
- Chromosome Location
- Chr. 12: 55973913 - 55996683 on Build GRCh38
- UniGene ID
- Hs.157659
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252036.1 |
NP_001238965.1
|
3, 4 |
488 |
|
NM_002868.3 |
NP_002859.1
|
3, 4 |
529 |
|
XM_005269051.2 |
XP_005269108.1
|
4, 5 |
550 |
|
XM_005269052.2 |
XP_005269109.1
|
3, 4 |
638 |
|
GenBank mRNA |
AF498937.1 |
- |
2, 3 |
308 |
|
AK292597.1 |
- |
3, 4 |
439 |
|
AK307468.1 |
- |
2, 3 |
200 |
|
BC032740.1 |
- |
3, 4 |
436 |
5538490 |
BC040143.1 |
- |
3, 4 |
466 |
|
BC050558.1 |
- |
3, 4 |
443 |
6191566 |
BC056422.1 |
- |
3, 4 |
418 |
6151079 |
BC065298.1 |
- |
3, 4 |
421 |
6146668 |
BX537408.1 |
- |
3, 4 |
463 |
|
CR536482.1 |
- |
2, 3 |
308 |
|
CR541690.1 |
- |
2, 3 |
308 |
|
X54871.1 |
- |
2, 3 |
328 |
|
View Full Product Details