Gene Transcripts

Gene Symbol
Pramel6
Entrez Gene ID
347711
Gene Name
preferentially expressed antigen in melanoma like 6
Gene Aliases
C80453, C80515
Chromosome Location
Chr. 2: 87506565 - 87510868 on Build GRCm38
UniGene ID
Mm.21815
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178249.2 NP_839980.1 3 1088
GenBank mRNA AF490342.1 - 3 1088
AK139830.1 - 4 1322

View Full Product Details