Gene Transcripts

Gene Symbol
Slc22a20
Entrez Gene ID
381203
Gene Name
solute carrier family 22 (organic anion transporter), member 20
Gene Aliases
Gm963, OAT1-L, mOAT6
Chromosome Location
Chr. 19: 5970232 - 5986551 on Build GRCm38
UniGene ID
Mm.329638
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198650.2 NP_941052.1 4 681
GenBank mRNA AK053009.1 - 1 2779
AY194118.1 - 4 680
BC046588.1 - 4 681 6309674

View Full Product Details