Gene Transcripts

Gene Symbol
Ube2u
Entrez Gene ID
381534
Gene Name
ubiquitin-conjugating enzyme E2U (putative)
Gene Aliases
Chromosome Location
Chr. 4: 100478867 - 100550145 on Build GRCm38
UniGene ID
Mm.318133
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033773.4 NP_001028945.2 6, 7 822
GenBank mRNA AK133379.1 - 5, 6 742
BC150880.1 - 6, 7 814 9087791
BC150888.1 - 6, 7 814 9087799
BC172031.1 - 6, 7 814 9054317

View Full Product Details