Gene Transcripts

Gene Symbol
Lmtk3
Entrez Gene ID
381983
Gene Name
lemur tyrosine kinase 3
Gene Aliases
BC059845, aatyk3
Chromosome Location
Chr. 7: 45783743 - 45806891 on Build GRCm38
UniGene ID
Mm.44928
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005511.3 NP_001005511.2 9, 10 943
NM_001290990.1 NP_001277919.1 8, 9 1098
GenBank mRNA AB288873.1 - 9, 10 937
BC057032.1 - 8, 9 831
BC083185.1 - 8, 9 1098 6855048
BC094377.1 - 9, 10 943 30536822

View Full Product Details