Gene Transcripts

Gene Symbol
Tas2r121
Entrez Gene ID
387349
Gene Name
taste receptor, type 2, member 121
Gene Aliases
T2R21, Tas2r13, Tas2r21, mGR21, mT2r48
Chromosome Location
Chr. 6: 132700090 - 132701007 on Build GRCm38
UniGene ID
Mm.377911
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207024.1 NP_996907.1 1 598
GenBank mRNA BC104410.1 - 1 598
BC104411.1 - 1 598 40050580

View Full Product Details