Gene Transcripts

Gene Symbol
Agr3
Entrez Gene ID
403205
Gene Name
anterior gradient 3
Gene Aliases
BCMP11, E030025L21Rik, Gm888
Chromosome Location
Chr. 12: 35925621 - 35949737 on Build GRCm38
UniGene ID
Mm.190508
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207531.3 NP_997414.2 7 552
GenBank mRNA AK053177.1 - 7 481
BC023499.1 - 7 553 5008294

View Full Product Details