Gene Transcripts

Gene Symbol
Ccdc36
Entrez Gene ID
434438
Gene Name
coiled-coil domain containing 36
Gene Aliases
Chromosome Location
Chr. 9: 108403502 - 108431841 on Build GRCm38
UniGene ID
Mm.425598
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135198.1 NP_001128670.1 7 1648
GenBank mRNA BC058624.1 - 7 1653

View Full Product Details