Gene Transcripts

Gene Symbol
Zcchc17
Entrez Gene ID
619605
Gene Name
zinc finger, CCHC domain containing 17
Gene Aliases
2810055E05Rik, HSPC251, LDC4, pNO40, pS1D
Chromosome Location
Chr. 4: 130315902 - 130359963 on Build GRCm38
UniGene ID
Mm.297196
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153160.4 NP_694800.1 4, 5 356
GenBank mRNA AJ272345.1 - 4, 5 312
AK012953.1 - 4, 5 370
AK152289.1 - 4, 5 357
AK161588.1 - 4, 5 351
AK168912.1 - 357
AY253297.1 - 4, 5 365
BC018382.1 - 3, 4 294 4215774
BC049231.1 - 4, 5 332 4161178

View Full Product Details