Gene Transcripts
- Gene Symbol
- Eif1
- Entrez Gene ID
- 20918
- Gene Name
- eukaryotic translation initiation factor 1
- Gene Aliases
- Sui1-rs1
- Chromosome Location
- Chr. 11: 100319994 - 100322096 on Build GRCm38
- UniGene ID
- Mm.13886
- Species
- Mus musculus
- Gene Symbol
- Nek7
- Entrez Gene ID
- 59125
- Gene Name
- NIMA (never in mitosis gene a)-related expressed kinase 7
- Gene Aliases
- 2810460C19Rik, AU020186
- Chromosome Location
- Chr. 1: 138484106 - 138619779 on Build GRCm38
- UniGene ID
- Mm.143817
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK165583.1 |
- |
|
2153 |
|
View Full Product Details